Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110505.1 | NP_001103975.1 | 4 | 587 | ||
NM_007446.2 | NP_031472.2 | 4 | 696 | |||
GenBank mRNA | AK029642.1 | - | 4 | 619 | ||
BC009121.1 | - | 4 | 570 | 3710941 | ||
CK791844.1 | - | 4 | 629 | |||
J00356.1 | - | 3 | 540 | |||
V00717.1 | - | 4 | 585 | |||
V00719.1 | - | 4 | 696 | |||
V00720.1 | - | 4 | 587 |