Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243043.1 | NP_001229972.1 | 3 | 540 | ||
NM_001243044.1 | NP_001229973.1 | 3 | 343 | |||
NM_007454.3 | NP_031480.2 | 3 | 343 | |||
GenBank mRNA | AK034140.1 | - | 3 | 543 | ||
AK155892.1 | - | 3 | 263 | |||
AK159273.1 | - | 3 | 234 | |||
AK160042.1 | - | 3 | 234 | |||
AK165747.1 | - | 3 | 259 | |||
AK170376.1 | - | 3 | 252 | |||
AK180383.1 | - | 3 | 136 | |||
BC008513.1 | - | 3 | 156 | 3601435 | ||
Y07919.1 | - | 3 | 198 |