Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap3d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007460.1 | NP_031486.1 | 2 | 375 | ||
GenBank mRNA | AB004305.1 | - | 2 | 375 | ||
AK141789.1 | - | 2 | 223 | |||
AK155518.1 | - | 2 | 425 | |||
AK155825.1 | - | 2 | 402 | |||
BC022171.1 | - | 2 | 380 | |||
BC037477.1 | - | 2 | 376 | |||
BC048786.1 | - | 2 | 365 | 6489789 | ||
BC053066.1 | - | 2 | 391 | 5704929 |