Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007473.4 | NP_031499.1 | 8 | 1291 | ||
GenBank mRNA | AB010100.1 | - | 8 | 1182 | ||
AB056091.1 | - | 8 | 1125 | |||
AK076642.1 | - | 8 | 1259 | |||
AK131744.1 | - | 8 | 1259 | |||
BC022223.1 | - | 6 | 921 | 4242417 |