Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TULP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007466.2 | NP_001007467.1 | 2, 3 | 1746 | ||
NM_020245.4 | NP_064630.2 | 2, 3 | 1746 | |||
XM_011535946.1 | XP_011534248.1 | 3, 4 | 2423 | |||
XM_017011063.1 | XP_016866552.1 | 4, 5 | 2572 | |||
XM_017011064.1 | XP_016866553.1 | 5, 6 | 2737 | |||
XM_017011065.1 | XP_016866554.1 | 4, 5 | 2490 | |||
XM_017011066.1 | XP_016866555.1 | 3, 4 | 2380 | |||
XM_017011067.1 | XP_016866556.1 | 3, 4 | 2473 | |||
XM_017011068.1 | XP_016866557.1 | 3, 4 | 2439 | |||
XM_017011069.1 | XP_016866558.1 | 4, 5 | 2575 | |||
XM_017011070.1 | XP_016866559.1 | 3, 4 | 2423 | |||
XM_017011071.1 | XP_016866560.1 | 4, 5 | 2575 | |||
GenBank mRNA | AB037818.1 | - | 2, 3 | 1746 | ||
AF219946.2 | - | 2, 3 | 1746 | |||
AF288480.1 | - | 2, 3 | 1746 | |||
AK304056.1 | - | 2, 3 | 961 | |||
BC152476.1 | - | 2, 3 | 1746 |