Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110204.1 | NP_001103674.1 | 6 | 823 | ||
NM_001110205.1 | NP_001103675.1 | 4 | 301 | |||
NM_007394.3 | NP_031420.2 | 5 | 578 | |||
GenBank mRNA | AF332087.1 | - | 3 | 211 | ||
AF332088.1 | - | 3 | 211 | |||
AK052246.1 | - | 413 | ||||
AK053428.1 | - | 6 | 822 | |||
AK150014.1 | - | 5 | 578 | |||
AK150075.1 | - | 5 | 578 | |||
BC058718.1 | - | 3 | 299 | 6418954 | ||
L15436.1 | - | 4 | 651 |