Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007400.2 | NP_031426.2 | 3, 4 | 485 | ||
GenBank mRNA | D50411.1 | - | 3, 4 | 485 |