Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007423.4 | NP_031449.3 | 3 | 258 | ||
GenBank mRNA | AK010934.1 | - | 3 | 234 | ||
AK075972.1 | - | 3 | 238 | |||
AK076053.1 | - | 3 | 234 | |||
AK076197.1 | - | 3 | 236 | |||
AK145975.1 | - | 3 | 234 | |||
AK146513.1 | - | 3 | 236 | |||
AK146524.1 | - | 3 | 236 | |||
AK167549.1 | - | 3 | 234 | |||
AK168426.1 | - | 3 | 236 | |||
AK168574.1 | - | 3 | 236 | |||
AK168576.1 | - | 3 | 234 | |||
AK168751.1 | - | 3 | 236 | |||
AK168818.1 | - | 2 | 112 | |||
AK168883.1 | - | 3 | 234 | |||
AK169046.1 | - | 3 | 236 | |||
AK169054.1 | - | 3 | 236 | |||
AK169147.1 | - | 3 | 234 | |||
BC066206.1 | - | 3 | 221 | 30469872 | ||
BY773400.1 | - | 3 | 265 | |||
V00743.1 | - | 3 | 232 |