Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001279349.1 | NP_001266278.1 | 1 | 443 | ||
NM_001279350.1 | NP_001266279.1 | 1 | 443 | |||
NM_020385.3 | NP_065118.2 | 1 | 443 | |||
NR_103995.1 | 1 | 443 | ||||
NR_103996.1 | 1 | 443 | ||||
GenBank mRNA | AF273304.1 | - | 1 | 391 | ||
AF295774.1 | - | 1 | 391 | |||
AK025493.1 | - | 1 | 297 | |||
AK304547.1 | - | 1 | 359 | |||
AK310164.1 | - | 1 | 346 | |||
AK314333.1 | - | 1 | 357 | |||
AK315904.1 | - | 2 | 294 | |||
AL136894.1 | - | 1 | 374 | |||
BC009274.2 | - | 1 | 346 | 3958331 | ||
BC071691.1 | - | 1 | 302 | |||
BX333016.2 | - | 1 | 333 | |||
DC387839.1 | - | 1 | 443 |