Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007467.3 | NP_031493.2 | 6 | 854 | ||
GenBank mRNA | AK154992.1 | - | 6 | 808 | ||
AK160871.1 | - | 6 | 806 | |||
AK170818.1 | - | 6 | 787 | |||
BC021877.1 | - | 6 | 833 | 4481838 | ||
BC106158.1 | - | 5 | 942 | |||
L04538.1 | - | 6 | 824 |