Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130408.1 | NP_001123880.1 | 3, 4 | 590 | ||
NM_007476.3 | NP_031502.1 | 3, 4 | 398 | |||
GenBank mRNA | AK077542.1 | - | 3, 4 | 348 | ||
AK148458.1 | - | 3, 4 | 362 | |||
AK148873.1 | - | 3, 4 | 352 | |||
AK150092.1 | - | 3, 4 | 371 | |||
AK150699.1 | - | 3, 4 | 348 | |||
AK151223.1 | - | 3, 4 | 348 | |||
AK151225.1 | - | 3, 4 | 371 | |||
AK151696.1 | - | 3, 4 | 324 | |||
AK151821.1 | - | 3, 4 | 346 | |||
AK152015.1 | - | 3, 4 | 344 | |||
AK152382.1 | - | 3, 4 | 346 | |||
AK152479.1 | - | 3, 4 | 605 | |||
AK153132.1 | - | 3, 4 | 365 | |||
AK159174.1 | - | 3, 4 | 348 | |||
AK167000.1 | - | 3, 4 | 327 | |||
AK167129.1 | - | 3, 4 | 361 | |||
AK167543.1 | - | 3, 4 | 348 | |||
AK167931.1 | - | 3, 4 | 362 | |||
AK168081.1 | - | 3, 4 | 535 | |||
AK168686.1 | - | 3, 4 | 348 | |||
AK169166.1 | - | 3, 4 | 346 | |||
AK170092.1 | - | 3, 4 | 350 | |||
AK170586.1 | - | 3, 4 | 323 | |||
AK193283.1 | - | 1, 2 | 57 | |||
BC021403.1 | - | 2, 3 | 341 | |||
BC031986.1 | - | 2, 3 | 315 | 4012689 | ||
D87898.1 | - | 3, 4 | 345 |