Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aatk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198785.1 | NP_001185714.1 | 14 | 4806 | ||
NM_001198787.1 | NP_001185716.1 | 14 | 4578 | |||
NM_007377.4 | NP_031403.2 | 14 | 5089 | |||
GenBank mRNA | AB093253.2 | - | 14 | 5089 | ||
AF011908.1 | - | 14 | 4545 | |||
AK036705.1 | - | 14 | 4580 | |||
AK076214.1 | - | 1 | 279 | |||
AK147239.1 | - | 10 | 5429 | |||
AK155100.1 | - | 14 | 4580 | |||
AY236858.1 | - | 14 | 4584 | |||
AY236859.1 | - | 14 | 4611 | |||
BC042837.1 | - | 13 | 5849 | |||
BC052323.1 | - | 13 | 5850 | |||
BC080846.1 | - | 14 | 4660 | 6856191 |