Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077190.2 | NP_001070658.1 | 11 | 2732 | ||
NM_001077192.2 | NP_001070660.1 | 9 | 2627 | |||
NM_001077193.2 | NP_001070661.1 | 8 | 2453 | |||
NM_007380.3 | NP_031406.2 | 10 | 2714 | |||
NM_145994.2 | NP_666106.1 | 10 | 2717 | |||
GenBank mRNA | AK034476.1 | - | 11 | 2614 | ||
AK152046.1 | - | 8 | 2565 | |||
AK152061.1 | - | 8 | 2317 | |||
AK152184.1 | - | 8 | 2317 | |||
AK153130.1 | - | 9 | 2535 | |||
AK164040.1 | - | 9 | 2543 | |||
AK167328.1 | - | 10 | 2673 | |||
AK167568.1 | - | 10 | 2672 | |||
AK167744.1 | - | 9 | 2574 | |||
AK191075.1 | - | 1 | 205 | |||
AY033645.1 | - | 9 | 2471 | |||
BC004657.1 | - | 10 | 2669 | 3498068 | ||
BC079642.1 | - | 10 | 2545 | 6816252 | ||
U17698.1 | - | 9 | 2362 |