Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK088287.1 | - | 1 | 166 | ||
AK145196.1 | - | 6 | 1471 | |||
AK145308.1 | - | 6 | 1470 | |||
AK151514.1 | - | 1 | 123 | |||
AK166349.1 | - | 6 | 1470 | |||
AK166498.1 | - | 6 | 1469 | |||
BC138611.1 | - | 6 | 1445 | 8861631 | ||
BC138614.1 | - | 6 | 1446 | 8861634 | ||
J04181.1 | - | 6 | 1452 | |||
X03672.1 | - | 6 | 1481 |