Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110204.1 | NP_001103674.1 | 9 | 1448 | ||
NM_001110205.1 | NP_001103675.1 | 7 | 926 | |||
NM_007394.3 | NP_031420.2 | 8 | 1203 | |||
GenBank mRNA | AF332087.1 | - | 6 | 836 | ||
AF332088.1 | - | 6 | 836 | |||
AK052246.1 | - | 1038 | ||||
AK053428.1 | - | 9 | 1447 | |||
AK150014.1 | - | 8 | 1203 | |||
AK150075.1 | - | 8 | 1203 | |||
BC058718.1 | - | 6 | 924 | 6418954 | ||
L15436.1 | - | 7 | 1276 |