Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272052.1 | NP_001258981.1 | 11 | 1167 | ||
NM_007398.4 | NP_031424.1 | 11 | 1167 | |||
GenBank mRNA | AF483480.1 | - | 11 | 1019 | ||
AF483481.1 | - | 11 | 1019 | |||
AK075899.1 | - | 11 | 1106 | |||
AK167489.1 | - | 11 | 1106 | |||
BC002075.1 | - | 11 | 1117 | 3484465 | ||
CT010224.1 | - | 11 | 1019 | |||
CT010255.1 | - | 11 | 1019 | |||
D19191.1 | - | 3 | 203 | |||
M10319.1 | - | 11 | 1089 |