Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007402.2 | NP_031428.2 | 2, 3 | 181 | ||
GenBank mRNA | AF013107.1 | - | 2, 3 | 167 | ||
AK136692.1 | - | 2, 3 | 181 |