Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089086.1 | - | 9 | 776 | ||
AK089169.1 | - | 9 | 798 | |||
AK150411.1 | - | 9 | 778 | |||
AK152675.1 | - | 9 | 795 | |||
AK152769.1 | - | 9 | 777 | |||
AK153065.1 | - | 9 | 777 | |||
AK155772.1 | - | 9 | 776 | |||
AK155912.1 | - | 9 | 796 | |||
AK160041.1 | - | 9 | 779 | |||
AK170019.1 | - | 9 | 801 | |||
AK171022.1 | - | 9 | 798 | |||
AK171060.1 | - | 9 | 776 | |||
BC025584.1 | - | 9 | 830 | 5324943 | ||
X13335.1 | - | 9 | 775 |