Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007432.2 | NP_031458.2 | 5 | 617 | ||
GenBank mRNA | BC115745.1 | - | 5 | 616 | ||
BC145627.1 | - | 3 | 813 |