Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007440.4 | NP_031466.2 | 14 | 2822 | ||
GenBank mRNA | AK087724.1 | - | 14 | 2824 |