Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | J00356.1 | - | 9 | 1374 | ||
V00717.1 | - | 10 | 1419 | |||
V00719.1 | - | 10 | 1530 | |||
V00720.1 | - | 10 | 1421 |