Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243043.1 | NP_001229972.1 | 7 | 1242 | ||
NM_001243044.1 | NP_001229973.1 | 7 | 1045 | |||
NM_007454.3 | NP_031480.2 | 7 | 1045 | |||
GenBank mRNA | AK034140.1 | - | 7 | 1245 | ||
AK155892.1 | - | 7 | 965 | |||
AK159273.1 | - | 7 | 936 | |||
AK160042.1 | - | 7 | 936 | |||
AK165747.1 | - | 7 | 962 | |||
AK170376.1 | - | 7 | 956 | |||
AK204765.1 | - | 1 | 16 | |||
BC008513.1 | - | 7 | 858 | 3601435 | ||
CA315828.1 | - | 2 | 265 | |||
Y07919.1 | - | 7 | 900 |