Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Astn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205204.1 | NP_001192133.1 | 8 | 1770 | ||
NM_007495.4 | NP_031521.2 | 8 | 1746 | |||
GenBank mRNA | AB093222.2 | - | 8 | 1332 | ||
AK045561.1 | - | 8 | 1501 | |||
AK138576.1 | - | 8 | 1515 | |||
AK147450.1 | - | 8 | 1516 | |||
AK164117.1 | - | 8 | 1735 | |||
AK165200.1 | - | 8 | 1499 | |||
BC054545.1 | - | 5 | 520 | |||
BC094666.1 | - | 8 | 1632 | 30545499 | ||
G49230.1 | - | 8 | 1356 | |||
U48797.1 | - | 8 | 1356 |