Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077593.1 | - | 2 | 315 | ||
AK141558.1 | - | 2 | 346 | |||
AK165931.1 | - | 2 | 312 | |||
BC050195.1 | - | 2 | 291 | 30010078 | ||
CD540305.1 | - | 3 | 441 | |||
U70859.1 | - | 3 | 449 |