Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bach2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109661.1 | NP_001103131.1 | 2 | 268 | ||
GenBank mRNA | AK162095.1 | - | 1 | 231 | ||
BC099420.1 | - | 1 | 185 | |||
BC131642.1 | - | 2 | 176 | 40109815 | ||
CJ092609.1 | - | 3 | 625 |