Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164090.1 | NP_001157562.1 | 4 | 384 | ||
NM_007530.3 | NP_031556.1 | 3 | 547 | |||
GenBank mRNA | AK090068.1 | - | 4 | 361 | ||
AK144754.1 | - | 4 | 358 | |||
AK218818.1 | - | 3 | 285 | |||
BC003303.1 | - | 3 | 451 | 3584161 | ||
BC021661.1 | - | 3 | 558 | 2646883 | ||
X78684.1 | - | 3 | 359 |