Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007542.4 | NP_031568.2 | 2 | 213 | ||
GenBank mRNA | AK086920.1 | - | 2 | 210 | ||
AK135496.1 | - | 2 | 214 | |||
AK155973.1 | - | 2 | 214 | |||
AK164867.1 | - | 2 | 214 | |||
AK171870.1 | - | 2 | 214 | |||
BC005452.1 | - | 2 | 207 | 3594469 | ||
BC019502.1 | - | 2 | 216 | 4237268 | ||
BC052857.1 | - | 2 | 198 | 30061850 | ||
BC057185.1 | - | 2 | 225 | |||
L20276.1 | - | 2 | 213 | |||
X53928.1 | - | 308 | ||||
Y11758.1 | - | 1 | 44 |