Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007557.3 | NP_031583.2 | 1 | 479 | ||
GenBank mRNA | AK020411.1 | - | 1 | 251 | ||
BC010771.1 | - | 1 | 451 | 4218495 | ||
CF540870.1 | - | 1 | 479 | |||
CJ094104.1 | - | 1 | 229 | |||
X56906.1 | - | 1 | 330 |