Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciita | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF042158.1 | - | 2 | 321 | ||
AF042159.1 | - | 3 | 627 | |||
AF100709.1 | - | 2 | 504 | |||
AF100710.1 | - | 2 | 220 | |||
AK040723.1 | - | 2 | 383 | |||
AK138822.1 | - | 2 | 413 | |||
AK143956.1 | - | 2 | 492 | |||
AK153609.1 | - | 2 | 317 | |||
AK155184.1 | - | 2 | 344 | |||
AK155311.1 | - | 2 | 413 | |||
BC107330.1 | - | 2 | 262 | |||
BC107331.1 | - | 2 | 262 | |||
U60653.1 | - | 2 | 281 |