Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007589.5 | NP_031615.1 | 3 | 298 | ||
GenBank mRNA | AK012247.1 | - | 3 | 226 | ||
AK012564.1 | - | 3 | 227 | |||
AK013068.1 | - | 3 | 220 | |||
AK152303.1 | - | 3 | 230 | |||
AK160636.1 | - | 3 | 233 | |||
AK161268.1 | - | 3 | 226 | |||
AK161984.1 | - | 3 | 227 | |||
AK168741.1 | - | 3 | 227 | |||
BC021347.1 | - | 3 | 229 | 4006686 | ||
BC051444.1 | - | 3 | 222 | 30072702 | ||
BC100301.1 | - | 3 | 213 | 30526959 | ||
BY095488.1 | - | 3 | 299 | |||
L31642.1 | - | 3 | 233 |