Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021629.3 | NP_067642.1 | 6 | 678 | ||
XM_005247692.2 | XP_005247749.1 | 6 | 599 | |||
XM_006713721.2 | XP_006713784.1 | 6 | 609 | |||
GenBank mRNA | AF300648.1 | - | 5 | 398 | ||
AK001890.1 | - | 6 | 678 | |||
AK290571.1 | - | 6 | 609 | |||
AK297174.1 | - | 6 | 475 | |||
AK310009.1 | - | 2 | 165 | |||
BC000873.1 | - | 6 | 488 | 3460570 |