Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164090.1 | NP_001157562.1 | 5 | 491 | ||
NM_007530.3 | NP_031556.1 | 4 | 654 | |||
GenBank mRNA | AK090068.1 | - | 5 | 468 | ||
AK144754.1 | - | 5 | 465 | |||
BC003303.1 | - | 4 | 558 | 3584161 | ||
BC021661.1 | - | 4 | 665 | 2646883 | ||
X78684.1 | - | 4 | 466 |