Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007545.2 | NP_031571.1 | 2 | 399 | ||
GenBank mRNA | BC128022.1 | - | 3 | 220 | ||
D83698.1 | - | 2 | 399 |