Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007549.2 | NP_031575.2 | 5 | 726 | ||
GenBank mRNA | AK089888.1 | - | 5 | 726 | ||
AK171640.1 | - | 5 | 673 | |||
BC030668.1 | - | 5 | 576 | 1178763 | ||
M30903.1 | - | 5 | 655 |