Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmpr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277216.1 | NP_001264145.1 | 5, 6 | 518 | ||
NM_001277217.1 | NP_001264146.1 | 5, 6 | 544 | |||
NM_001277218.1 | NP_001264147.1 | 4, 5 | 557 | |||
NM_001277220.1 | NP_001264149.1 | 5, 6 | 692 | |||
NM_007560.4 | NP_031586.1 | 4, 5 | 711 | |||
GenBank mRNA | AK086130.1 | - | 5, 6 | 504 | ||
AK140158.1 | - | 5, 6 | 616 | |||
AK162844.1 | - | 4, 5 | 392 | |||
BC065106.1 | - | 4, 5 | 492 | 30357130 | ||
BC065143.1 | - | 4, 5 | 698 | 30629345 | ||
BC138638.1 | - | 4, 5 | 438 | 8861658 | ||
BC138640.1 | - | 4, 5 | 438 | 8861660 | ||
CV557650.1 | - | 4, 5 | 555 | |||
Z23143.1 | - | 5, 6 | 536 |