Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007497.3 | NP_031523.3 | 7 | 1915 | ||
GenBank mRNA | AK042385.1 | - | 1 | 182 | ||
AK148500.1 | - | 31 | ||||
BC006871.1 | - | 8 | 1819 | 2646620 | ||
M63725.1 | - | 6 | 1704 |