Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007504.2 | NP_031530.2 | 23 | 3397 | ||
GenBank mRNA | AK132537.1 | - | 8 | 1114 | ||
AY081946.1 | - | 23 | 3428 | |||
BC036292.1 | - | 23 | 3397 | 4191741 | ||
X67140.1 | - | 7 | 810 |