Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007505.2 | NP_031531.1 | 12 | 2192 | ||
GenBank mRNA | AK043976.1 | - | 12 | 1961 | ||
AK076572.1 | - | 12 | 1960 | |||
AK098155.1 | - | 12 | 1970 | |||
AK150426.1 | - | 12 | 1963 | |||
AK151128.1 | - | 12 | 1969 | |||
AK151920.1 | - | 12 | 1964 | |||
AK152054.1 | - | 12 | 1971 | |||
AK152890.1 | - | 12 | 1963 | |||
AK153167.1 | - | 12 | 1964 | |||
AK153282.1 | - | 12 | 1960 | |||
AK159540.1 | - | 12 | 1960 | |||
AK159758.1 | - | 12 | 1963 | |||
AK164110.1 | - | 12 | 1963 | |||
AK164193.1 | - | 10 | 1666 | |||
AK166709.1 | - | 12 | 1963 | |||
AK167863.1 | - | 12 | 1940 | |||
AK168198.1 | - | 12 | 1970 | |||
AK168369.1 | - | 12 | 1962 | |||
AK168617.1 | - | 12 | 1960 | |||
AK168879.1 | - | 12 | 1960 | |||
AK168890.1 | - | 12 | 1960 | |||
AK168932.1 | - | 12 | 1960 | |||
AK169084.1 | - | 12 | 1963 | |||
AK169300.1 | - | 12 | 1960 | |||
AK169308.1 | - | 12 | 1965 | |||
AK169414.1 | - | 12 | 1963 | |||
AK204538.1 | - | 1 | 24 | |||
BC014854.1 | - | 12 | 1947 | 4500893 | ||
BY465601.1 | - | 1 | 192 | |||
L01062.1 | - | 12 | 1947 |