Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007510.2 | NP_031536.2 | 5 | 385 | ||
GenBank mRNA | AB074758.1 | - | 5 | 387 | ||
AK010603.1 | - | 5 | 402 | |||
AK139791.1 | - | 5 | 402 | |||
AK146162.1 | - | 5 | 399 | |||
AK149356.1 | - | 5 | 421 | |||
AK167644.1 | - | 5 | 344 | |||
AK169854.1 | - | 5 | 399 | |||
AK186366.1 | - | 5 | 337 | |||
AK189499.1 | - | 5 | 334 | |||
AK189578.1 | - | 5 | 337 | |||
BC003421.1 | - | 5 | 385 | 2598896 | ||
BC055438.1 | - | 5 | 419 | 3995744 |