Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301424.1 | NP_001288353.1 | 10 | 1628 | ||
NM_007513.4 | NP_031539.3 | 10 | 1744 | |||
GenBank mRNA | AK139342.1 | - | 10 | 1720 | ||
AK147991.1 | - | 10 | 1653 | |||
BC068234.1 | - | 10 | 1635 | |||
BC145779.1 | - | 9 | 1549 | 40131082 | ||
BC145781.1 | - | 9 | 1549 | 40131084 | ||
JN226407.1 | - | 8 | 1464 | |||
JN226408.1 | - | 8 | 1464 | |||
JN226410.1 | - | 8 | 1464 | |||
M26687.1 | - | 10 | 1662 |