Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bach2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109661.1 | NP_001103131.1 | 5 | 3052 | ||
GenBank mRNA | BC051242.1 | - | 3 | 1096 | ||
BC131642.1 | - | 5 | 2960 | 40109815 |