Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285453.1 | NP_001272382.1 | 4 | 643 | ||
NM_007522.3 | NP_031548.1 | 4 | 1160 | |||
GenBank mRNA | AK151792.1 | - | 4 | 608 | ||
AK169002.1 | - | 4 | 605 | |||
BC006762.1 | - | 4 | 1128 | 3257589 | ||
L37296.1 | - | 4 | 1168 |