Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007537.1 | NP_031563.1 | 4 | 2303 | ||
GenBank mRNA | AF030769.1 | - | 4 | 2303 | ||
AK004680.1 | - | 4 | 2333 | |||
AK172925.1 | - | 2 | 2131 | |||
AY170344.3 | - | 2284 | ||||
BC040369.1 | - | 2 | 2794 | 4511215 |