Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opn1sw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007538.3 | NP_031564.1 | 5 | 2023 | ||
GenBank mRNA | BC026021.1 | - | 5 | 1990 | 4511808 |
Set Membership: |
G-protein coupled receptor |