Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007549.2 | NP_031575.2 | 11 | 1558 | ||
GenBank mRNA | AK089888.1 | - | 11 | 1558 | ||
AK171640.1 | - | 11 | 1505 | |||
BC030668.1 | - | 11 | 1408 | 1178763 | ||
CT010240.1 | - | 10 | 1138 | |||
M30903.1 | - | 11 | 1487 |