Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007553.3 | NP_031579.2 | 3 | 1786 | ||
GenBank mRNA | AK132433.1 | - | 3 | 1751 | ||
AK133923.1 | - | 3 | 1786 | |||
AK161862.1 | - | 3 | 1590 | |||
BC100344.1 | - | 3 | 746 | 30846365 |