Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007556.2 | NP_031582.1 | 7 | 1946 | ||
GenBank mRNA | AK014797.1 | - | 484 | |||
AK041210.1 | - | 2304 | ||||
AK135770.1 | - | 7 | 1579 | |||
BC138593.1 | - | 7 | 1866 | 8861613 | ||
BC138595.1 | - | 7 | 1866 | 8861615 | ||
J04566.1 | - | 7 | 1528 | |||
X80992.1 | - | 7 | 1945 |