Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007568.5 | NP_031594.1 | 6 | 1705 | ||
GenBank mRNA | AK046658.1 | - | 5 | 2986 | ||
AK050206.1 | - | 6 | 1521 | |||
AK076272.1 | - | 6 | 1625 |