Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007585.3 | NP_031611.1 | 6, 7 | 526 | ||
GenBank mRNA | AK012563.1 | - | 7, 8 | 528 | ||
AK088645.1 | - | 6, 7 | 515 | |||
AK146096.1 | - | 6, 7 | 514 | |||
AK150586.1 | - | 6, 7 | 514 | |||
AK152561.1 | - | 6, 7 | 514 | |||
AK160746.1 | - | 6, 7 | 514 | |||
AK164507.1 | - | 6, 7 | 519 | |||
AK165888.1 | - | 6, 7 | 514 | |||
AK167894.1 | - | 6, 7 | 513 | |||
AK168083.1 | - | 6, 7 | 514 | |||
BC003327.1 | - | 6, 7 | 526 | 3156634 | ||
BC004659.1 | - | 2, 3 | 1422 | 3497991 | ||
BC005763.1 | - | 6, 7 | 500 | 2655513 | ||
D10024.1 | - | 5, 6 | 459 | |||
M14044.1 | - | 5, 6 | 459 |