Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007594.4 | NP_031620.1 | 2 | 192 | ||
NM_184053.3 | NP_908942.1 | 2 | 192 | |||
GenBank mRNA | AK135295.1 | - | 2 | 117 | ||
AK147756.1 | - | 2 | 145 | |||
AK148178.1 | - | 2 | 145 | |||
AK160795.1 | - | 2 | 145 | |||
AK161807.1 | - | 2 | 149 | |||
AK163674.1 | - | 2 | 180 | |||
AK166705.1 | - | 2 | 145 | |||
AY225334.1 | - | 2 | 118 | |||
BC051423.1 | - | 2 | 153 | 3989906 | ||
CJ084197.1 | - | 2 | 195 | |||
U81829.1 | - | 2 | 118 |